Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tris base buffer, powder, Corning®

Tris base buffer, powder, Corning®

Tris base buffer, powder, 1.000 g
Catalog # MDTC61-233-RR
Supplier:   Corning
Tris base buffer, powder, Corning®
Catalog # MDTC61-233-RR
Supplier:  Corning
CAS Number:  
Tris base buffer, powder, Corning®
Tris base buffer, powder, Corning®

Tris base buffer, powder, 1.000 g

CAS Number:  
Price
277.00 SFr.
Log in for availability
1 * 1.000 g
Product Details & Documents

Tris is commonly used in buffer solutions in biochemical and molecular biological research.

This primary amine has an effective pH range of 7,0 to 9,2 and is reported to inhibit a number of different enzymes, limiting its usefulness in protein studies to some extent. This tris base buffer is provided in powder form.

Specifications

Specifications

  • Pk:
    1.000 g
  • Description:
    Tris base buffer, powder
  • Cat. No.:
    MDTC61-233-RR
  • Storage temperature:
    Ambient
  • Shipping temperature:
    Ambient

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.