Tris base buffer, powder, Corning®
Tris base buffer, powder, 5.000 g
Supplier: Corning
Jump to:
Product Details & Documents
Tris is commonly used in buffer solutions in biochemical and molecular biological research.
This primary amine has an effective pH range of 7,0 to 9,2 and is reported to inhibit a number of different enzymes, limiting its usefulness in protein studies to some extent. This tris base buffer is provided in powder form.
Documents
Specifications
Specifications
- Pk:5.000 g
- Description:Tris base buffer, powder
- Cat. No.:MDTC61-233-RT
- Storage temperature:Ambient
- Shipping temperature:Ambient