Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Corning® Trypsin

Corning® Trypsin

100 ml, 6
Catalog # 25-052-CI
Supplier:   Corning
Corning® Trypsin
Catalog # 25-052-CI
Supplier:  Corning
CAS Number:  
Corning® Trypsin
Corning® Trypsin

100 ml, 6

CAS Number:  
Price
81.60 SFr.
Log in for availability
1 * 6 ST
Product Details & Documents

0,05% Trypsin/0,53 mM EDTA in HBSS (Hank’s Balanced Salt Solution) with sodium bicarbonate, without calcium and magnesium.

  • Trypsin EDTA 1X 0,05% Trypsin/0,53 mM EDTA in HBSS with sodium bicarbonate, without calcium and magnesium
  • Porcine Parvovirus tested

Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging. Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances the efficacy of trypsin. This product contains trypsin at a concentration of 0,05% with 0,53 mM EDTA, and does not contain sodium bicarbonate.

Specifications

Specifications

  • Pk:
    6
  • Pk Info:
    100 ml
  • Cat. No.:
    25-052-CI
  • Enzyme name:
    Trypsin
  • CAS No.:
    9002-07-7

Specifications