ContactUsLinkComponent
FLUORESCENT DYE 700-I PHALLOIDIN FLUORESCENCE DYE 700-I WITH PHALLOIDIN CONJUGATE. IN COMPARISON WITH ALEXA FLUOR 700, FLUORESCENCE DYE 700-I ARE BRIGHTER WITH STRONGER ABSORPTION AT 633NM. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 700-I IS WELL SE 1 * 300 Assays
Catalog # ABNOU0302
Supplier:  Abnova
FLUORESCENT DYE 700-I PHALLOIDIN FLUORESCENCE DYE 700-I WITH PHALLOIDIN CONJUGATE. IN COMPARISON WITH ALEXA FLUOR 700, FLUORESCENCE DYE 700-I ARE BRIGHTER WITH STRONGER ABSORPTION AT 633NM. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 700-I IS WELL SE 1 * 300 Assays
Catalog # ABNOU0302
Supplier:  Abnova