CUP REPLACEMENT FOR VORTEX 1 1 * 1 items
Catalog # IKAA3382051
Supplier: IKA
Frequently Bought Together
MEDELA EINWEG SEKRETBEUTEL 2.5 LITER MIT GELIERGRANULAT 1 * 30 ST
Supplier: MARKET SOURCE PART PROCESS
Boiling stones, 30g 1 * 1 items
Supplier: VELP SCIENTIFIC
NEW POWER SUPPLY FOR THE AIR SAMPLER MD8 1 * 1 items
Supplier: Sartorius
Gas sensor 0-20% CO2 Steri max. 190 C 1 * 1 items
Supplier: MEMMERT
INOX BASE PLATE SECURA 1 * 1 items
Supplier: Sartorius Balances
GAS TUBING FOR PENSKY-MARTENS 1:2 SETA 1 * 1 items
Supplier: STANHOPE SETA
Set of clips (20 pieces) 1 * 20 items
Supplier: BUCHI
Spare element for 240V Bibby D4000 water still (Bibby D240H) 1 * 1 items
Supplier: Cole Parmer
Accessories for Research® Plus pipettes
Supplier: EPPENDORF
Speed sensor 1 * 1 items
Supplier: Cole Parmer
DELIVERY 1 * 1 items
Supplier: Thermo Scientific
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach