Bagrack 3500 ml -2 compartments, dimensions 35.5 x 20.5 x 20 cm. 1 * 1 items
Catalog # INSC221352
Supplier: INTERSCIENCE
Bagrack 3500 ml -2 compartments, dimensions 35.5 x 20.5 x 20 cm. 1 * 1 items
Catalog # INSC221352
Supplier: INTERSCIENCE
Frequently Bought Together
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Square Bottles, PET
Supplier: LAB3 OWN PRODUCTION
VWR®, Cryo coder cap inserts
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
InfinityLab Well Plates
Supplier: AGILENT
Optical bottom plates, 96- and 384-well, Nunc™
Supplier: Thermo Fisher Scientific
Polypropylene centrifuge tubes, PerformR®
Supplier: LABCON
Closures, PPCO, natural, with colour coder, 11 mm
Supplier: Thermo Fisher Scientific
Cryoboxes, 133×133 mm
Supplier: RATIOLAB
Eppendorf® DNA LoBind Plates
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
96-Well clear polystyrene microplates, Corning®
Supplier: Corning
Fibronectin-coated flasks, Corning® BioCoat®
Supplier: Corning
Nunc™ EasYFlask™ Flasks with Nunclon™ Supra Surface
Supplier: Thermo Fisher Scientific
Cryogenic tubes
Supplier: TPP
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
Supplier: LAB3 OWN PRODUCTION