Polymer-based SEC column for both aqueous and organic solvents for the analysis of proteins, hormones, polyelectrolytes and surfactants. Exclusion limit 9.000 Da (pullulan standard) for low molecular weight substances. 1 * 1 items
Catalog # SHOWF7600000
Supplier: Resonac
Frequently Bought Together