ContactUsLinkComponent
Petri Dishes 90 x 13,5mm “SPACE & MEDIA SAVER” with 3 vents & ISO mark, printable. Features smaller diameter, lower height and unique recessed lid permitting space savings in incubator, refrigerator and during shipping. The surface all around the bas 1 * 624 items
Catalog # PHOE110
Supplier:  PHOENIX BIOMEDICAL
Petri Dishes 90 x 13,5mm “SPACE & MEDIA SAVER” with 3 vents & ISO mark, printable. Features smaller diameter, lower height and unique recessed lid permitting space savings in incubator, refrigerator and during shipping. The surface all around the bas 1 * 624 items
Catalog # PHOE110
Supplier:  PHOENIX BIOMEDICAL
Frequently Bought Together