ContactUsLinkComponent
Two-lens magnifier – It is composed of two lenses whose diameter is 22 mm. First lens magnification: 4x second lens magnification: 6x total magnification: 10x. 1 * 1 items
Katalognr. OPTK4986
Supplier:  OPTIKA
Two-lens magnifier – It is composed of two lenses whose diameter is 22 mm. First lens magnification: 4x second lens magnification: 6x total magnification: 10x. 1 * 1 items
Katalognr. OPTK4986
Supplier:  OPTIKA
Frequently Bought Together