ContactUsLinkComponent
Counting glass with millimetric scale - 5x counting glass with double millimetric ruler for measurement. 1 * 1 items
Catalog # OPTK4090
Supplier:  OPTIKA
Counting glass with millimetric scale - 5x counting glass with double millimetric ruler for measurement. 1 * 1 items
Catalog # OPTK4090
Supplier:  OPTIKA
Frequently Bought Together