ContactUsLinkComponent
Centimeter-graduated magnifier for insects - With ventilation holes in order to observe alive insects. With double 2x - 4x lens. Chamber dimensions: Ø 70 mm, height 75 mm. 1 * 1 items
Catalog # OPTK4026
Supplier:  OPTIKA
Centimeter-graduated magnifier for insects - With ventilation holes in order to observe alive insects. With double 2x - 4x lens. Chamber dimensions: Ø 70 mm, height 75 mm. 1 * 1 items
Catalog # OPTK4026
Supplier:  OPTIKA
Frequently Bought Together