VWR®, Racks for VLSB Shaking Baths
Stainless steel test tube rack for 12 and 18 l baths, 119, 0,5 ml
Catalog # 462-0509
Supplier: VWR Collection
Jump to:
Product Details & Documents
Documents
VWR® for baths, thermostats and circulators
Product Literature
Specifications
- For tubes:0,5 ml
- No. of Holes:119
- Description:Stainless steel test tube rack for 12 and 18 l baths
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



