Screw caps
Caps, PP, white, LDPE foam lined, 48-400
Catalog # EPSC067-4800WBP
Supplier: EP SCIENTIFIC
Jump to:
Product Details & Documents
LDPE-lined white caps.
- For general-purpose HDPE bulk separate containers
- Same high quality caps used on certified HDPE environmental sample containers
Ordering information: Bottles sold separately (IR013-series and IR012-series).
Specifications
- Thread:48-400
- Description:Caps, PP, white, LDPE foam lined
- Seq:04
- Cat. No.:EPSC067-4800WBP
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact