ContactUsLinkComponent
Centrifuge 5430 New capacity pack EU-plug includes centrifuge 5430 230V, rotors FA-45-48-11 and FA-45-16-17, 200 Eppendorf tubes 5,0ml and 2 white racks for 16 5,0ml tubes 1 * 1 KIT
Catalog # 521-1884
Supplier:  EPPENDORF
Centrifuge 5430 New capacity pack EU-plug includes centrifuge 5430 230V, rotors FA-45-48-11 and FA-45-16-17, 200 Eppendorf tubes 5,0ml and 2 white racks for 16 5,0ml tubes 1 * 1 KIT
Catalog # 521-1884
Supplier:  EPPENDORF
Frequently Bought Together