ContactUsLinkComponent
CoolRack XT 5 mL: holds 12 x 5 mL centrifuge tubes. Maintains temperature through direct contact with ice, dry ice, water, liquid nitrogen or other common laboratory temperature sources. Use for benchtop cooling, (snap)freezing or heating/thawing sam 1 * 1 items
Catalog # BCISBCS-539
Supplier:  BIOCISION
CoolRack XT 5 mL: holds 12 x 5 mL centrifuge tubes. Maintains temperature through direct contact with ice, dry ice, water, liquid nitrogen or other common laboratory temperature sources. Use for benchtop cooling, (snap)freezing or heating/thawing sam 1 * 1 items
Catalog # BCISBCS-539
Supplier:  BIOCISION
Frequently Bought Together