ContactUsLinkComponent
BioCision™ XT Starter PCR96 System for ice-free PCR/qPCR sample preparation. Low profile, open platform ice-free cooler holds one 96- well PCR plate, strip-wells or 100 µl tubes 1 * 1 items
Catalog # BCISBCS-556
Supplier:  BIOCISION
undefined
BioCision™ XT Starter PCR96 System for ice-free PCR/qPCR sample preparation. Low profile, open platform ice-free cooler holds one 96- well PCR plate, strip-wells or 100 µl tubes 1 * 1 items
Catalog # BCISBCS-556
Supplier:  BIOCISION
Frequently Bought Together