ContactUsLinkComponent
SecinH3 is a selective inhibitor of cytohesins, blocking human, mouse, and Drosophila cytohesins with IC50 values of 2.4 to 5.6 µM.{ 14565} It binds the Sec7 domain of these small Arf-GEFs but does not bind Sec7 domains of larger Arf-GEFs.{25739} In 1 * 5 mg
Catalog # CAYM10009570-5
Supplier:  Cayman Chemical
SecinH3 is a selective inhibitor of cytohesins, blocking human, mouse, and Drosophila cytohesins with IC50 values of 2.4 to 5.6 µM.{ 14565} It binds the Sec7 domain of these small Arf-GEFs but does not bind Sec7 domains of larger Arf-GEFs.{25739} In 1 * 5 mg
Catalog # CAYM10009570-5
Supplier:  Cayman Chemical
Frequently Bought Together