Microscope slides, SuperFrost®, Menzel Gläser
Rose, Unwrapped
Catalog # 630-2861
Supplier: Epredia
Jump to:
Product Details & Documents
These slides are made of extra-white glass, washed clean and polished.
- Labelling area resistant to all common solvents
- Six colours for identification and colour coding
- Thick labelling area to prevent slides from sticking together or getting scratched
Suitable for in vitro diagnostic (IVD) applications.
Complies with ISO 8037/I
Certifications: ISO 8037/I.
Packaging: In cardboard boxes of 50.
Documents
Specifications
- Packed:Unwrapped
- Colour:Rose
- Seq:309