Dialysis tubing Trial kit Spectra/Por 7 pre-treated standard regenerated cellulose (RC) membrane MWCO 8kD 32mm flat width (20,4mm di ameter) 3,3ml/cm including tubing closures and openers wet 1 * 1 m
Catalog # SPEC132115T
Supplier: Spectrum Laboratories
Frequently Bought Together