Order Entry
Switzerland
Orders LinkContactUsLinkComponent
pH Buffers

pH Buffers

pH buffer, 3,9 M NaCl, 9,21, 6×250 ml, 6
Catalog # METN52004110
Supplier:   Mettler - Toledo
pH Buffers
Catalog # METN52004110
Supplier:  Mettler - Toledo
CAS Number:  
pH Buffers

pH buffer, 3,9 M NaCl, 9,21, 6×250 ml, 6

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 6 ST
Product Details & Documents
Specifications

Specifications

  • Pk:
    6
  • Description:
    pH buffer, 3,9 M NaCl
  • Pk Info:
    6×250 ml
  • pH value:
    9,21

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.