HUMIDITY STANDARD 75%RH WITH SCS CERT. 1 * 5 Ampoul
Catalog # ROTREA75-SCS
Supplier: ROTRONIC INSTRUMENTS
HUMIDITY STANDARD 75%RH WITH SCS CERT. 1 * 5 Ampoul
Catalog # ROTREA75-SCS
Supplier: ROTRONIC INSTRUMENTS
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Mixed Anions Standard, 7 components in H₂O
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Disodium oxalate, Certipur®, Supelco®
Supplier: Merck
Hydrochloric acid 0.01 mol/l (0.01 N) Reag. Ph. Eur. 1043504
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions
Supplier: SI Analytics
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
ICP-MS CAL STD 4 1 * 1 items
Supplier: PerkinElmer
Conductivity standards
Supplier: Hach
Simethicone, Reference Standard
Supplier: USP