HUMIDITY STANDARD 11,3%RH WITH SCS CERT. 1 * 5 Ampoul
ROTREA11-SCS
Frequently Bought Together
pH buffers, ready to use
: WTW
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
Humidity standards
: ROTRONIC INSTRUMENTS
Buffer solutions in sachets
: Mettler - Toledo
VWR®, Conductivity standard solutions
: VWR Chemicals