Microscope slides, SuperFrost®, Menzel Gläser
Green, Unwrapped
Catalog # 630-2863
Supplier: Epredia
Jump to:
Product Details & Documents
These slides are made of extra-white glass, washed clean and polished.
- Labelling area resistant to all common solvents
- Six colours for identification and colour coding
- Thick labelling area to prevent slides from sticking together or getting scratched
Suitable for in vitro diagnostic (IVD) applications.
Complies with ISO 8037/I
Zertifizierung: ISO 8037/I.
Verpackung: In cardboard boxes of 50.
Dokumente
MENZAA00008032E011_TDS_EN MENZAG00008532E01_TDS_EN
Technisches Datenblatt
Specifications
- Packed:Unwrapped
- Colour:Green
- Seq:315