BIS-TRIS, 0,2 M Buffer Solutions
BIS-TRIS buffer solution, 0,2 M, 7,0, Glass bottle, 100 mL
CAS No.: 6976-37-0
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00002853
- Concentration:0,2 M
- Pack type:Glass bottle
- Pk:100 mL
- Description:BIS-TRIS buffer solution
- pH value:7,0
- Cat. No.:J63346.AE
- Odour/scent:Odourless
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00002853
- Appearance:Colourless
- CAS No.:6976-37-0
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



