Orders LinkContactUsLinkComponent
Trypsin
Trypsin 0,25% EDTA in HBSS, without calcium, without magnesium, with phenol red, 500 ml
N° de catalogue L0931-500
Fournisseur:   Biowest
Trypsin
N° de catalogue L0931-500
Fournisseur:  Biowest
Numéro CAS:  
Trypsin
Trypsin
Trypsin 0,25% EDTA in HBSS, without calcium, without magnesium, with phenol red, 500 ml
Numéro CAS:  
Prix
SFr. 64.70
Connexion Pour la disponibilité
1 * 500 mL
Product Details & Documents
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
Spécifications
Spécifications
  • Pk:
    500 ml
  • Description:
    Trypsin 0,25% EDTA in HBSS, without calcium, without magnesium, with phenol red
Spécifications