Trypsin
Trypsin 0,25% EDTA in HBSS, without calcium, without magnesium, with phenol red, 100 ml
Nome del fornitore: Biowest
Vai a
Product Details & Documents
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
Documents
Specifiche
Specifiche
- Pk:100 ml
- Description:Trypsin 0,25% EDTA in HBSS, without calcium, without magnesium, with phenol red
Specifiche
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



