Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Amphotericin B

Amphotericin B

20 mL
Catalog # L0009-020
Supplier:   Biowest
Amphotericin B
Catalog # L0009-020
Supplier:  Biowest
CAS Number:  1397-89-3
Amphotericin B
Amphotericin B

20 mL

CAS Number:  1397-89-3
Price
SFr. 16.10
Log in for availability
1 * 20 mL
Product Details & Documents

Amphotericin B used in tissue culture to prevent growth of yeasts and fungi. It changes the function and integrity of eucaryotic cell membranes by forming complexes with sterols (cholesterol) hence having no effect on bacteria. Cause leakage of glucose.

  • Yellow coloured, with a pH of 10 – 11,5
  • Shelf life of 24 month

Interferes with fungal membrane permeability by forming channels in the membranes and causing small molecules to leak out.

Caution: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

Synonyms:  (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
  • CAS Number:1397-89-3
  • Formula:C₄₇H₇₃NO₁₇
  • MDL Number:MFCD00877763
  • Molecular Weight:924.08 g/mol

Documents

Specifications

Specifications

  • Pk:
    20 mL

Specifications