ContactUsLinkComponent
Cutter, megasafe, trapezoid blade, rounded tip, length:153mm, width:26mm, height:38mm, cutting depth:20.1mm, basic material:metal. 1 * 1 items
Catalog # MARO116006.02
Supplier:  MARTOR ARGENTAX
Cutter, megasafe, trapezoid blade, rounded tip, length:153mm, width:26mm, height:38mm, cutting depth:20.1mm, basic material:metal. 1 * 1 items
Catalog # MARO116006.02
Supplier:  MARTOR ARGENTAX
Frequently Bought Together