I-TAC Human Recombinant (Interferon-inducible T-cell alpha chemoattractant) produced in E.coli is a single, non-glycosylated, polype ptide chain containing 73 amino acids and having a molecular mass of 8300 Dalton. The I-TAC is purified by p 1 * 20 µG
Catalog # BIRBORB80138-20
Supplier: Biorbyt
Frequently Bought Together