ContactUsLinkComponent
TFF-1 Human Recombinant produced in E.coli is homodimer, non-glycosylated, polypeptide chain containing x 60 amino acids which inclu des 40 amino acid trefoil motif containing conserved interamolecular disulfide bonds and having total molecu 1 * 20 µG
Catalog # BIRBORB80104-20
Supplier:  Biorbyt
TFF-1 Human Recombinant produced in E.coli is homodimer, non-glycosylated, polypeptide chain containing x 60 amino acids which inclu des 40 amino acid trefoil motif containing conserved interamolecular disulfide bonds and having total molecu 1 * 20 µG
Catalog # BIRBORB80104-20
Supplier:  Biorbyt