ContactUsLinkComponent
Betacellulin Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 80 amino acids and hav ing a molecular mass of 9 kDa. Betacellulin Human Recombinant is purified by proprietary chromatographic tec 1 * 5 µG
Catalog # BIRBORB80011-5
Supplier:  Biorbyt
Betacellulin Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 80 amino acids and hav ing a molecular mass of 9 kDa. Betacellulin Human Recombinant is purified by proprietary chromatographic tec 1 * 5 µG
Catalog # BIRBORB80011-5
Supplier:  Biorbyt