ContactUsLinkComponent
Recombinant Dengue Virus Subtype Envelope 32kDa, produced in E.coli, suitable for Gold Conjugation, strong Dengue IgM reactive teste d by lateral flow device. 1 * 100 µG
Catalog # BIRBORB81815-100
Supplier:  Biorbyt
Recombinant Dengue Virus Subtype Envelope 32kDa, produced in E.coli, suitable for Gold Conjugation, strong Dengue IgM reactive teste d by lateral flow device. 1 * 100 µG
Catalog # BIRBORB81815-100
Supplier:  Biorbyt