ContactUsLinkComponent
Angiostatin Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 259 amino acids and havi ng a molecular mass of approximately 30.0 kDa. The Ang K1-3 is purified by proprietary chromatographic techn 1 * 20 µG
Catalog # BIRBORB80910-20
Supplier:  Biorbyt
Angiostatin Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 259 amino acids and havi ng a molecular mass of approximately 30.0 kDa. The Ang K1-3 is purified by proprietary chromatographic techn 1 * 20 µG
Catalog # BIRBORB80910-20
Supplier:  Biorbyt