ContactUsLinkComponent
Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza virus, strain B/Florida/04/06. The Influenza Virus was pu rified by Ultracentrifugation with 10-40 sucrose gradient. 1 * 50 µG
Catalog # BIRBORB81794-50
Supplier:  Biorbyt
Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza virus, strain B/Florida/04/06. The Influenza Virus was pu rified by Ultracentrifugation with 10-40 sucrose gradient. 1 * 50 µG
Catalog # BIRBORB81794-50
Supplier:  Biorbyt