ContactUsLinkComponent
Recombinant Human Trypsin-2 is free from any animal and human sources. Recombinant Human Trypsin-2 expressed in E.coli having Mw of 24kDa is purified by standard chromatography techniques. Recombinant Human Trypsin-2 is free from foreign en 1 * 1 mg
Catalog # BIRBORB81439-1
Supplier:  Biorbyt
Recombinant Human Trypsin-2 is free from any animal and human sources. Recombinant Human Trypsin-2 expressed in E.coli having Mw of 24kDa is purified by standard chromatography techniques. Recombinant Human Trypsin-2 is free from foreign en 1 * 1 mg
Catalog # BIRBORB81439-1
Supplier:  Biorbyt