ContactUsLinkComponent
Antide acetate (Ac-AA10-NH2)is an LHRH antagonist and represses LH and FSH release from the pituitary gland. Antide has molecular fo rmula of C82-H108-N17-O14 cl1, a.a. sequence of Ac-{D-2-Nal}-{D4 cl-Phe}{D-4-pAL)-Ser-Lys-Lys-Leu-Lys-Pro-{D 1 * 5 mg
Catalog # BIRBORB80701-5
Supplier:  Biorbyt
Antide acetate (Ac-AA10-NH2)is an LHRH antagonist and represses LH and FSH release from the pituitary gland. Antide has molecular fo rmula of C82-H108-N17-O14 cl1, a.a. sequence of Ac-{D-2-Nal}-{D4 cl-Phe}{D-4-pAL)-Ser-Lys-Lys-Leu-Lys-Pro-{D 1 * 5 mg
Catalog # BIRBORB80701-5
Supplier:  Biorbyt