ContactUsLinkComponent
Mucosae-associated Epithelial Chemokine (MEC)/CCL28 (CC chemokine ligand 28) is a secreted CC chemokineexpressed primarily by epithe lial cells of the bronchioles, salivary gland, mammary gland and colon. MEC signals throughthe CCR10 recepto 1 * 20 µG
Catalog # BIRBORB107783-20
Supplier:  Biorbyt
Mucosae-associated Epithelial Chemokine (MEC)/CCL28 (CC chemokine ligand 28) is a secreted CC chemokineexpressed primarily by epithe lial cells of the bronchioles, salivary gland, mammary gland and colon. MEC signals throughthe CCR10 recepto 1 * 20 µG
Catalog # BIRBORB107783-20
Supplier:  Biorbyt