ContactUsLinkComponent
Material: low density polyethylene (LDPE) Kenylene CP1060 resin: additive-free, very low outgassing Color: Transparent For clean rooms Thickness: 150 µm Width: 4 (101,6mm) Packing: Roll of 500' (150 m) in double-packaging 1 * 1 items
Catalog # KNFCLT106K0400
Supplier:  KNF CLEANROOM
Material: low density polyethylene (LDPE) Kenylene CP1060 resin: additive-free, very low outgassing Color: Transparent For clean rooms Thickness: 150 µm Width: 4 (101,6mm) Packing: Roll of 500' (150 m) in double-packaging 1 * 1 items
Catalog # KNFCLT106K0400
Supplier:  KNF CLEANROOM
Frequently Bought Together