ContactUsLinkComponent
Aa s/chamber organic, mk7 complete 1 /pk for use with organic solvents. includes mixing paddle,neb. block with integral nebulizer, p ress.relief bung, liquid trap and drain tube (with integral float) 1 * 1 items
Catalog # VARI110866890
Supplier:  VARIAN
Aa s/chamber organic, mk7 complete 1 /pk for use with organic solvents. includes mixing paddle,neb. block with integral nebulizer, p ress.relief bung, liquid trap and drain tube (with integral float) 1 * 1 items
Catalog # VARI110866890
Supplier:  VARIAN
Frequently Bought Together