ContactUsLinkComponent
Fiehn gc/ms metabolomics standards kit contains 4 ampoules d27 myristic acids 1 ampoule ea: fames/d27-myristic acids, pyridine and m stfa/ 1% tmcs 1 * 1 items
Catalog # VARI400505
Supplier:  VARIAN
Fiehn gc/ms metabolomics standards kit contains 4 ampoules d27 myristic acids 1 ampoule ea: fames/d27-myristic acids, pyridine and m stfa/ 1% tmcs 1 * 1 items
Catalog # VARI400505
Supplier:  VARIAN
Frequently Bought Together