ContactUsLinkComponent
All-glass sample intro system radial icp. includes radial torch with ball joint, glass transfer tube, double pass glass cyclonic spr aychamber and adjustable mount 1 * 1 items
Catalog # VARI9910117900
Supplier:  VARIAN
All-glass sample intro system radial icp. includes radial torch with ball joint, glass transfer tube, double pass glass cyclonic spr aychamber and adjustable mount 1 * 1 items
Catalog # VARI9910117900
Supplier:  VARIAN
Frequently Bought Together