Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Polysorbate 20, J.T.Baker®

Glass bottle, amber, narrow mouth, 500 mL
Catalog # X251-07
Supplier:   Avantor
Polysorbate 20, J.T.Baker®
Catalog # X251-07
Supplier:  Avantor
CAS Number:  9005-64-5
Polysorbate 20, J.T.Baker®
Polysorbate 20, J.T.Baker®

Glass bottle, amber, narrow mouth, 500 mL

CAS Number:  9005-64-5
Price
SFr. 380.00
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Density:1.1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:500-018-3

 

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Identification (by IR):
    Passes Test

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    500 mL

Specifications