ContactUsLinkComponent
C18Q x 3µm particle size x 100Å pore size, 150mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C183-03N03S
C18Q x 3µm particle size x 100Å pore size, 150mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C183-03N03S
Frequently Bought Together