ContactUsLinkComponent
C18Q x 3µm particle size x 100Å pore size, 100mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C183-03M02S
C18Q x 3µm particle size x 100Å pore size, 100mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C183-03M02S
Frequently Bought Together