ContactUsLinkComponent
C18G x 3µm particle size x 200Å pore size, 100mm length x 1.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C181-03M01S
C18G x 3µm particle size x 200Å pore size, 100mm length x 1.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C181-03M01S
Frequently Bought Together