ContactUsLinkComponent
C18P x 5µm particle size x 120Å pore size, 50mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Katalognr. SGEA2POL-05L03S
C18P x 5µm particle size x 120Å pore size, 50mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Katalognr. SGEA2POL-05L03S
Frequently Bought Together