ContactUsLinkComponent
C18P x 5µm particle size x 120Å pore size, 50mm length x 3.0mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2POL-05L03P
C18P x 5µm particle size x 120Å pore size, 50mm length x 3.0mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2POL-05L03P
Frequently Bought Together