ContactUsLinkComponent
C18P x 3µm particle size x 120Å pore size, 150mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2POL-03N02S
C18P x 3µm particle size x 120Å pore size, 150mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2POL-03N02S
Frequently Bought Together