ContactUsLinkComponent
PFP x 5µm particle size x 120Å pore size, 100mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2PFP-05M46S
undefined
PFP x 5µm particle size x 120Å pore size, 100mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2PFP-05M46S
Frequently Bought Together