ContactUsLinkComponent
C18G x 3µm particle size x 120Å pore size, 150mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C185-03N02S
C18G x 3µm particle size x 120Å pore size, 150mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C185-03N02S
Frequently Bought Together